Product Image
TPP Techno Plastic Products™
Duotest™ pH Paper Roll
Featuring two indicator zones for more accurate readings. Macherey Nagel™ Duotest™ pH Paper Rolls give the user the best possible accuracy in pH measurement.
Have Questions?
Catalog NumberpH Range
90301TS
also known as 90301
1 to 12
Catalog number 90301TS
also known as 90301
Price (USD)
-
pH Range:
1 to 12
Featuring two indicator zones for more accurate readings. Macherey Nagel™ Duotest™ pH Paper Rolls give the user the best possible accuracy in pH measurement.
  • Provides rapid and reliable results
  • Available for a range of pH gradations
  • Shows two different colours for each pH-value at intervals of 0.3 to 1 pH unit
  • Allows more accurate reading and good estimation of intermediate values
Specifications
FormatRoll
Graduations1, 2, 3, 4, 5, 6, 7, 8, 9, 10, 11, 12
Quantity1 Each
TypepH Paper
Width (Metric)10 mm
pH Range1 to 12
Unit SizeCase of 5