Provides for the quick and easy determination of lead hydrogen sulphide in samples. Macherey Nagel™ Lead Acetate Test Paper is toxic and needs to be declared as hazardous.
Have Questions?
Catalog Number
Color Indicator
90744TSPK
also known as 90744
White to Brown/Black
Catalog number 90744TSPK
also known as 90744
Price (USD)
-
Color Indicator:
White to Brown/Black
Provides for the quick and easy determination of lead hydrogen sulphide in samples. Macherey Nagel™ Lead Acetate Test Paper is toxic and needs to be declared as hazardous.
Qualitative
Sulfide containing solutions also give a positive reaction
Non-toxic and safe sulfide test paper is an alternative
Specifications
Color IndicatorWhite to Brown/Black
Dimensions (L x W)5000 x 7 mm
For Use With (Application)Determination of lead hydrogen sulphide