Product Image
3M Australia

Chlorohexidine Gluconate 4% Antimicrobial Surgical Scrub 10 pcs, 10 x 500 mL Bottles with Pump Pack - Not TGA Approved Not Allowed For Clinical Use

Chlorohexidine Gluconate 4% Antimicrobial Surgical Scrub 10 pcs, 10 x 500 mL Bottles with Pump Pack - Not TGA Approved Not Allowed For Clinical Use
Have Questions?
Catalog number 9241-P
Price (USD)
-
Chlorohexidine Gluconate 4% Antimicrobial Surgical Scrub 10 pcs, 10 x 500 mL Bottles with Pump Pack - Not TGA Approved Not Allowed For Clinical Use

3M Chlorohexidine Gluconate 4% hand scrub 10pcs 10 x 500mL bottles with pump pack Broad spectrum antimicrobial surgical scrub Not TGA Approved Not allowed for Clinical Use

Specifications
Composition4% w/w Chlorhexidine Gluconate
Volume (Metric)500 mL
SterilityNon-sterile
pH4 to 7
Unit SizeEach