Specifications
Boiling Point2850°C
CAS1305-62-0
Chemical Name or MaterialCalcium hydroxide
IUPAC Namecalcium dihydroxide
InChI KeyAXCZMVOFGPJBDE-UHFFFAOYSA-L
MDL NumberMFCD00010901
Melting Point580°C
Molecular FormulaCaH2O2
PackagingPlastic
SMILES[OH-].[OH-].[Ca++]
GradeLaboratory
Molecular Weight (g/mol)74.09
Quantity1 kg
Unit SizeEach