KingFisher™ Pure DNA Blood Kit
Thermo Scientific™
KingFisher™ Pure DNA Blood Kit
Achieve rapid, reproducible purification of high-quality DNA from fresh or frozen whole blood treated with EDTA or citrate with this unique nucleic acid purification workflow.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
9801019696 Samples
98010496384 Samples
Catalog number 98010196
Price (USD)
-
Quantity:
96 Samples
Achieve rapid, reproducible purification of high-quality DNA. The Thermo Scientific™ KingFisher™ Pure DNA Blood Kit, together with a KingFisher magnetic particle processor, allows rapid and reproducible purification of high-quality DNA from fresh or frozen whole blood treated with EDTA or citrate. Thermo Scientific KingFisher Kits complete the unique nucleic acid purification workflow providing an optimized high throughput method for extreme flexibility.
Features
  • Produces isolated DNA that is free of inhibitors and ready for use in many downstream applications
  • Easy-to-use protocols for safe sample-handling, as well as minimal user input, maximizing walkaway time.
  • Excellent performance and consistency of results optimized to produce high yields of quality DNA
  • Large volume sample input possible up to 3mL of blood per sample
  • Flexible throughput from one sample to up to 96 samples per run
  • Minimized hands-on time increases efficiency
  • Compatible with most common downstream analysis and applications
Recommended for:
Genomics, target identification, veterinary assays, biomarker discovery, quality control
Specifications
Elution Volume200μL sample: 100μL (Duo) or 150μL (Flex). 1mL sample: 500μL. Recommended but adjustable
Quantity96 Samples
Sample TypeFresh or Frozen Whole Blood Treated with EDTA or Citrate
Sample Volume200 μL or 1 mL
Test Time40 min. (200 μL), 45 min. (1 mL)
Unit SizeEach
Frequently asked questions (FAQs)
Are the KingFisher Pure kits discontinued?
All KingFisher Pure kits are discontinued. We recommend using the MagMAX kits which are optimized for use with the KingFisher instruments. Please contact our Technical Support team at techsupport@thermofisher.com to find out which MagMAX kit we recommend as an alternative to the KingFisher Pure Kits.
Find additional tips, troubleshooting help, and resources within our Automated Nucleic Acid Purification Support Center.
What alternatives do you recommend for the discontinued KingFisher Pure kits?
Please contact our Technical Support team at techsupport@thermofisher.com to find out which MagMAX kits we recommend as an alternative to the KingFisher Pure Kits.
Find additional tips, troubleshooting help, and resources within our Automated Nucleic Acid Purification Support Center.