Indiko™ and Konelab™ System Albumin to CRP Specific Protein Reagents
Indiko™ and Konelab™ System Albumin to CRP Specific Protein Reagents
Actual product may vary
Thermo Scientific™

Indiko™ and Konelab™ System Albumin to CRP Specific Protein Reagents

Increase testing efficiency with our ready-to-use, barcoded Thermo Scientific™ Indiko™ and Konelab™ System Albumin to CRP Specific Protein Reagents.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
981932Complement C4; Kit Size: 2 x 3mL antiserum
981251CRP Control; Kit size: 5 x 1mL
981674CRP Calibration Set; Kit size: 5 x 1mL
981679CRP Control High; Kit size: 5 x 1mL
981699CRP; Kit Size: 2 x 3mL antiserum
981798CRP High Sensitivity; Kit Size: 1 x 4mL antiserum
981852CRP High Sensitivity Control; Kit size: 3 x 1mL
981877Albumin U Calibrator; Kit Size: 2 x 2mL
981878Albumin U Control; Kit size: 2 x 2mL
981879Albumin U Control High; Kit size: 2 x 2mL
981921APO A1/APO B Calibrator; Kit size: 2 x 1mL
981927Albumin MST; Kit Size: 2 x 3mL antiserum
981929Apolipoprotein A1; Kit Size: 2 x 3mL antiserum
981930Apolipoprotein B; Kit Size: 2 x 3mL antiserum
981933CRP; Kit Size: 1 x 3mL antiserum
981931Complement C3; Kit Size: 2 x 3mL antiserum
Catalog number 981932
Price (USD)
-
Description:
Complement C4; Kit Size: 2 x 3mL antiserum
Receive best-in-class methodologies and standardized performance with Thermo Scientific™ Indiko™ and Konelab™ System Specific Protein Reagents, which include a specific protein test menu of 15 immunoturbidimetric methods. All reagents are ready to go and packaged in volume-optimized, barcoded kits; in-house evaluation as well as optimization of reagents guarantees continuous support and optimum results.

All Indiko and Konelab methods are based on carefully-designed protocols for barcoded system reagents. Calibrators and controls are available in separate kits.

Highlights:

  • High-quality reagents are barcoded for automatic identification
  • Most reagents are delivered in ready-to-use liquid form
  • Special attention has been paid to the optimization of package sizes
  • The CE marked products are compliant with EU regulatory requirements for in vitro diagnostic medical devices

Ordering Information: Not available in the U.S.A.

Specific Protein Applications for Indiko and Konelab Analyzers:

  • Albumin / Microalbuminuria
  • Apolipoprotein A1 & B
  • Complement C3 & C4
  • C-reactive protein
  • D-Dimer
  • Ferritin
  • HbA1c
  • IgA, G & M
  • Rheumatoid factors
  • Soluble transferrin receptor
  • Transferrin
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
DescriptionComplement C4; Kit Size: 2 x 3mL antiserum
Quantity2 x 3 mL
Sample TypeSerum
TypeReagent
Detectable AnalytesComplement C4
FormLiquid
Unit SizeEach