Acetonitrile, Certified, for HPLC-RMN with Low Propionitrile Level
Acetonitrile, Certified, for HPLC-RMN with Low Propionitrile Level
Acetonitrile, Certified, for HPLC-RMN with Low Propionitrile Level
Fisher Chemical™

Acetonitrile, Certified, for HPLC-RMN with Low Propionitrile Level

Have Questions?
Catalog NumberQuantity
A-0636-17
also known as A/0636/17
2.5 L
Catalog number A-0636-17
also known as A/0636/17
Price (USD)
-
Quantity:
2.5 L
Request bulk or custom format
Specifications
Boiling Point81°C
CAS75-05-8
ColorColorless
Formula Weight41.05g/mol
IUPAC Nameacetonitrile
View more
Acetonitrile, Certified, for HPLC-RMN with Low Propionitrile Level