Thermo Scientific Chemicals

Mesitaldehyde, 97%, Thermo Scientific Chemicals

Catalog number: A10104.09
10 g, Each
Thermo Scientific Chemicals

Mesitaldehyde, 97%, Thermo Scientific Chemicals

Catalog number: A10104.09
10 g, Each
Quantity
Catalog number: A10104.09
also known as A10104-09
Price (USD)

Chemical Identifiers

CAS
487-68-3
Form
Liquid
Comment
May contain trace amounts of particulates matter from the manufacturing process
Identification (FTIR)
Conforms
Free acid (titration)
≤1.0%
Refractive Index
1.5515-1.5565 @ 20?C

Description

Mesitaldehyde is used in organic synthesis, and also used as medicine and other fine chemical products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Mesitaldehyde is used in organic synthesis, and also used as medicine and other fine chemical products.

Solubility
Soluble in Chloroform

Notes
Air Sensitive. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text