Thermo Scientific Chemicals

2,6-Dichloroindophenol sodium salt hydrate, Thermo Scientific Chemicals

Catalog number: A10107.06
5 g, Each
Thermo Scientific Chemicals

2,6-Dichloroindophenol sodium salt hydrate, Thermo Scientific Chemicals

Catalog number: A10107.06
5 g, Each
Quantity
Catalog number: A10107.06
also known as A10107-06
Price (USD)

Chemical Identifiers

CAS
1266615-56-8
IUPAC Name
sodium 4-[(3,5-dichloro-4-oxocyclohexa-2,5-dien-1-ylidene)amino]benzen-1-olate
Molecular Formula
C12H6Cl2NNaO2
InChI Key
CVSUAFOWIXUYQA-UHFFFAOYSA-M
SMILES
[Na+].[O-]C1=CC=C(C=C1)N=C1C=C(Cl)C(=O)C(Cl)=C1
Appearance (Color)
Dark green to black
Comment
Suitable for ascorbic acid determination
Specific Absorbance (E 1%/1cm)
≥700 @ lambda max
lambda-max (UV-Vis)
638-645nm (50% ethanol + 2ml 1M sodium carbonate soln)
Form
Powder

Description

2,6-Dichloroindophenol sodium salt hydrate is suitable for vitamin C determination. It acts as a redox dye to investigate the efficiency of gold nanoparticles (Au-NP) for the enzymatic activity of glucose oxidase. It is also used to compose UVB specific dosimeter and to measure the rate of photosynthesis. It serves as a good reducing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Dichloroindophenol sodium salt hydrate is suitable for vitamin C determination. It acts as a redox dye to investigate the efficiency of gold nanoparticles (Au-NP) for the enzymatic activity of glucose oxidase. It is also used to compose UVB specific dosimeter and to measure the rate of photosynthesis. It serves as a good reducing agent.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text