Thermo Scientific Chemicals

Allyl phenyl sulfone, 98%, Thermo Scientific Chemicals

Catalog number: A10401.06
5 g, Each
Thermo Scientific Chemicals

Allyl phenyl sulfone, 98%, Thermo Scientific Chemicals

Catalog number: A10401.06
5 g, Each
Quantity
Catalog number: A10401.06
also known as A10401-06
Price (USD)

Chemical Identifiers

CAS
16212-05-8
IUPAC Name
(prop-2-ene-1-sulfonyl)benzene
Molecular Formula
C9H10O2S
InChI Key
KYPIULIVYSQNNT-UHFFFAOYSA-N
SMILES
C=CCS(=O)(=O)C1=CC=CC=C1
Refractive Index
1.5455-1.5495 @ 20?C
Appearance (Color)
Clear colorless to yellow
Assay (GC)
≥97.5%
Form
Liquid
Identification (FTIR)
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text