Thermo Scientific Chemicals

Salicylaldehyde azine, 97%, Thermo Scientific Chemicals

Catalog number: A10527.09
10 g, Each
Thermo Scientific Chemicals

Salicylaldehyde azine, 97%, Thermo Scientific Chemicals

Catalog number: A10527.09
10 g, Each
Quantity

Chemical Identifiers

CAS
959-36-4
IUPAC Name
(6E)-6-[(2-{[(1E)-6-oxocyclohexa-2,4-dien-1-ylidene]methyl}hydrazin-1-yl)methylidene]cyclohexa-2,4-dien-1-one
Molecular Formula
C14H12N2O2
InChI Key
SPEXYYIULCBQJR-WGDLNXRISA-N
SMILES
O=C1C=CC=C\C1=C/NN\C=C1/C=CC=CC1=O
Appearance (Color)
Yellow
Assay (HPLC)
≥96.0%
Melting Point (clear melt)
214.0-224.0?C
Form
Crystals or powder or crystalline powder or lumps
Identification (FTIR)
Conforms

Description

Salicylaldehyde azine is used in the preparation of 2-oxo-2H-1-benzopyran-3-carboxylic acid 2-(2-oxo-2H-1-benzopyran-3-carbonyl)hydrazide by reacting with propadienedione . It has anthelmintic activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Salicylaldehyde azine is used in the preparation of 2-oxo-2H-1-benzopyran-3-carboxylic acid 2-(2-oxo-2H-1-benzopyran-3-carbonyl)hydrazide by reacting with propadienedione . It has anthelmintic activity.

Solubility
Soluble in dimethyl sulfoxide, methanol and ethyl acetate. Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text