Thermo Scientific Chemicals

Methyl diethylphosphonoacetate, 97%, Thermo Scientific Chemicals

Catalog number: A10644.14
25 g, Each
Thermo Scientific Chemicals

Methyl diethylphosphonoacetate, 97%, Thermo Scientific Chemicals

Catalog number: A10644.14
25 g, Each
Quantity
Catalog number: A10644.14
also known as A10644-14
Price (USD)

Chemical Identifiers

CAS
1067-74-9
IUPAC Name
methyl 2-(diethoxyphosphoryl)acetate
Molecular Formula
C7H15O5P
InChI Key
CTSAXXHOGZNKJR-UHFFFAOYSA-N
SMILES
CCOP(=O)(CC(=O)OC)OCC
Appearance (Color)
Clear colorless
Assay (GC)
≥96.0%
Refractive Index
1.4300-1.4350 @ 20?C
Form
Liquid

Description

Methyl diethylphosphonoacetate is used as a reagent in the preparation of allylic fluorides compounds and in regioselective Diels-Alder reactions. It also serves as a reactant in the preparation of substituted thiophenes and furans, which finds application in type 2 diabetes treatment. Further, it acts as a precursor in the synthesis of pyridone alkaloids and immunosuppressive cyclopentanediol derivatives. In addition to this, it is utilized in the modification of botulinum neurotoxin serotype A protease inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl diethylphosphonoacetate is used as a reagent in the preparation of allylic fluorides compounds and in regioselective Diels-Alder reactions. It also serves as a reactant in the preparation of substituted thiophenes and furans, which finds application in type 2 diabetes treatment. Further, it acts as a precursor in the synthesis of pyridone alkaloids and immunosuppressive cyclopentanediol derivatives. In addition to this, it is utilized in the modification of botulinum neurotoxin serotype A protease inhibitors.

Solubility
Miscible with tetrahydrofuran.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text