2-Bromo-3'-methoxyacetophenone, 98%
2-Bromo-3'-methoxyacetophenone, 98%
2-Bromo-3'-methoxyacetophenone, 98%
Thermo Scientific Chemicals

2-Bromo-3'-methoxyacetophenone, 98%

CAS: 5000-65-7 | C9H9BrO2 | 229.073 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A10716.14
also known as A10716-14
Price (USD)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS5000-65-7
IUPAC Name2-bromo-1-(3-methoxyphenyl)ethan-1-one
Molecular FormulaC9H9BrO2
InChI KeyIOOHBIFQNQQUFI-UHFFFAOYSA-N
SMILESCOC1=CC=CC(=C1)C(=O)CBr
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Melting Point (clear melt)60.0-67.0?C
Appearance (Color)Pale cream
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only