Thermo Scientific Chemicals

3-Chlorophenylacetic acid, 98+%, Thermo Scientific Chemicals

Catalog number: A11210.06
5 g, Each
Thermo Scientific Chemicals

3-Chlorophenylacetic acid, 98+%, Thermo Scientific Chemicals

Catalog number: A11210.06
5 g, Each
Quantity
Catalog number: A11210.06
also known as A11210-06
Price (USD)

Chemical Identifiers

CAS
1878-65-5
IUPAC Name
2-(3-chlorophenyl)acetic acid
Molecular Formula
C8H7ClO2
InChI Key
WFPMUFXQDKMVCO-UHFFFAOYSA-N
SMILES
OC(=O)CC1=CC=CC(Cl)=C1
Appearance (Color)
White to cream
Form
Crystals or powder or crystalline powder or flakes or chunks
Assay (HPLC)
≥98.0%
Melting Point (clear melt)
75.5-81.5?C
Assay (Aqueous acid-base Titration)
≥98.0 to ≤102.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text