Thermo Scientific Chemicals

4-Bromo-o-xylene, 97%, Thermo Scientific Chemicals

Catalog number: A14736.06
5 g, Each
Thermo Scientific Chemicals

4-Bromo-o-xylene, 97%, Thermo Scientific Chemicals

Catalog number: A14736.06
5 g, Each
Quantity
Catalog number: A14736.06
also known as A14736-06
Price (USD)

Chemical Identifiers

CAS
583-71-1
IUPAC Name
4-bromo-1,2-dimethylbenzene
Molecular Formula
C8H9Br
InChI Key
QOGHRLGTXVMRLM-UHFFFAOYSA-N
SMILES
CC1=CC=C(Br)C=C1C
Appearance (Color)
Clear colorless
Refractive Index
1.5535-1.5585 @ 20?C
Assay (GC)
≥96.0%
Form
Liquid

Description

4-Bromo-o-xylene is used as synthetic organic materials and soluble polyimide resin intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromo-o-xylene is used as synthetic organic materials and soluble polyimide resin intermediates.

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text