Thermo Scientific Chemicals

Indole-5-carboxylic acid, 98%, Thermo Scientific Chemicals

Catalog number: A18627.14
25 g, Each
Thermo Scientific Chemicals

Indole-5-carboxylic acid, 98%, Thermo Scientific Chemicals

Catalog number: A18627.14
25 g, Each
Quantity
Catalog number: A18627.14
also known as A18627-14
Price (USD)

Chemical Identifiers

CAS
1670-81-1
IUPAC Name
1H-indole-5-carboxylate
Molecular Formula
C9H6NO2
InChI Key
IENZCGNHSIMFJE-UHFFFAOYSA-M
SMILES
[O-]C(=O)C1=CC=C2NC=CC2=C1
Assay (HPLC)
≥97.5%
Appearance (Color)
White to cream to yellow
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%
Form
Powder
Melting Point (clear melt)
208.0-214.0?C

Description

Indole-5-carboxylic acid is the suitable reagent used to study the intermolecular excited state proton transfer in indole-2-carboxylic acid and indole-5-carboxylic acid in various solvents in acidic, basic, and neutral media by steady state and time resolved fluorescence spectroscopy. It may be used in the electrochemical synthesis of poly(indole-5-carboxylic acid) (PICA) films. Also used as reactant for preparation of tryptophan dioxygenase inhibitors pyridyl-ethenyl-indoles, as potential anticancer immunomodulators, synthesis of indirubin derivatives and amide conjugates with ketoprofen, as inhibitors of Gli1-mediated transcription in Hedgehog pathway.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Indole-5-carboxylic acid is the suitable reagent used to study the intermolecular excited state proton transfer in indole-2-carboxylic acid and indole-5-carboxylic acid in various solvents in acidic, basic, and neutral media by steady state and time resolved fluorescence spectroscopy. It may be used in the electrochemical synthesis of poly(indole-5-carboxylic acid) (PICA) films. Also used as reactant for preparation of tryptophan dioxygenase inhibitors pyridyl-ethenyl-indoles, as potential anticancer immunomodulators, synthesis of indirubin derivatives and amide conjugates with ketoprofen, as inhibitors of Gli1-mediated transcription in Hedgehog pathway.

Solubility
Soluble in ethanol (50 mg/ml), dimethyl sulfoxide and methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text