Product Image
Invitrogen™
Attune™ CytPix™ Violet 6 Conversion Kit, for red laser
The Attune CytPix Violet 6 Conversion Kit is designed to reconfigure Attune NxT four-channel violet instruments to allow detection ofRead more
Have Questions?
Catalog NumberQuantity
A509281 Kit
Catalog number A50928
Price (USD)
-
Quantity:
1 Kit
The Attune CytPix Violet 6 Conversion Kit is designed to reconfigure Attune NxT four-channel violet instruments to allow detection of six channels of violet. A conversion kit is required for each original and any upgraded lasers of the instrument. The conversion is carried out by a Field Service Engineer.
Specifications
For Use With (Application)Flow Cytometry
For Use With (Equipment)Attune CytPix Flow Cytometer
IncludesFilters, dichroics
Product LineDJM
Quantity1 Kit
Regulatory StatusRUO
Shipping ConditionRT
Unit SizeEach