Specifications
Boiling Point117°C
CAS64-19-7
Chemical Name or MaterialAcetic Acid
IUPAC Nameacetic acid
InChI KeyQTBSBXVTEAMEQO-UHFFFAOYSA-N
MDL NumberMFCD00036152
Melting Point16°C
Molecular FormulaC2H4O2
PackagingPlastic
SMILESCC(O)=O
SynonymEthanoic acid
GradeLaboratory
Molecular Weight (g/mol)60.05
Quantity20 L
Unit SizeEach