Thermo Scientific Chemicals

Allylchlorodimethylsilane, 94%, Thermo Scientific Chemicals

Catalog number: B22237.09
10 g, Each
Thermo Scientific Chemicals

Allylchlorodimethylsilane, 94%, Thermo Scientific Chemicals

Catalog number: B22237.09
10 g, Each
Quantity
Catalog number: B22237.09
also known as B22237-09
Price (USD)

Chemical Identifiers

CAS
4028-23-3
IUPAC Name
chlorodimethyl(prop-2-en-1-yl)silane
Molecular Formula
C5H11ClSi
InChI Key
KMVZWUQHMJAWSY-UHFFFAOYSA-N
SMILES
C[Si](C)(Cl)CC=C
Appearance (Color)
Clear colorless
Identification (FTIR)
Conforms
Refractive Index
1.4260-1.4320 @ 20?C
Form
Liquid
Assay (GC)
≥92.0%

Description

Allylchlorodimethylsilane is used in the synthesis of cyclo-1,1?,4,4?-bis(1,1,3,3-tetramethyl-1,3-disiloxanediyl)dibenzene, silylating agent for steroids for analysis by GC-MS. Also used in the preparation of phenylenebis(silanediyl triflates), useful synthons for organosilicon polymers, O-silylation of phenyl sulfones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Allylchlorodimethylsilane is used in the synthesis of cyclo-1,1′,4,4′-bis(1,1,3,3-tetramethyl-1,3-disiloxanediyl)dibenzene, silylating agent for steroids for analysis by GC-MS. Also used in the preparation of phenylenebis(silanediyl triflates), useful synthons for organosilicon polymers, O-silylation of phenyl sulfones.

Solubility
Soluble in most organic solvents; reacts with alcohols, amines, and water.

Notes
Moisture sensitive. Store away from strong oxidizing agents, strong bases, water/ moisture.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text