Oxoid™ Prepared Yeast Extract Agar Bottled Media, 498-502 g, 500 mL sirop bottle, 10 bottles
Thermo Scientific™

Oxoid™ Prepared Yeast Extract Agar Bottled Media, 498-502 g, 500 mL sirop bottle, 10 bottles

Enumerate microorganisms in potable and freshwater samples with this agar.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFill WeightFormat
BO0635M100.0 to 101.5 g125 mL sirop bottle
BO0556G20 to 20.3 gUniversal Bottle
BO0556T250 to 254 g300 mL Sirop Bottle
BO0556V498 to 502 g500 mL Sirop Bottle
EB0556F15.0 to 15.3 gUniversal Bottle
Catalog number BO0635M
Price (USD)
-
Fill Weight:
100.0 to 101.5 g
Format:
125 mL sirop bottle
Yeast Extract Agar is used for the enumeration of microorganisms in potable and freshwater samples.
Specifications
ColorStraw
Fill Weight100.0 to 101.5 g
Format125 mL sirop bottle
FormulationYeast extract (3.0 g/L); Peptone (5 g/L); Agar (15 g/L)
Packaging Type10 sirop bottles with screw cap closures in a box.
Quantity10 x 100 mL
Shelf Life52 weeks
Manufacturing OriginUnited Kingdom
Product TypeAgar
Unit SizeEach
Contents & Storage
2 to 25°C