Tryptone Soya Broth
Thermo Scientific™

Tryptone Soya Broth

A non-selective broth with neutralizers.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFill WeightFormatShelf LifeQuantity
BO1108J90 to 91.5 g125 mL Sirop Bottle26 weeks10/Pk.
BO0479Z90-91.5 g 125 mL sirop bottle 26 weeks 10 x 90 mL
BO1023J90 to 91.5 g125 mL Sirop Bottle52 weeks10/Pk.
BO1084D9.0 to 9.3 gUniversal Bottle26 weeks24/Pk.
Catalog number BO1108J
Price (USD)
-
Fill Weight:
90 to 91.5 g
Format:
125 mL Sirop Bottle
Shelf Life:
26 weeks
Quantity:
10/Pk.
Tryptone Soya Broth is a non-selective broth with neutralizers.
Specifications
ColorStraw
Fill Weight90 to 91.5 g
Format125 mL Sirop Bottle
Packaging Type10 Sirop Bottles with ROPP cap closures/Box
Quantity10/Pk.
Shelf Life26 weeks
Manufacturing OriginUnited Kingdom
Product TypeMicrobiology Media
Unit SizeEach
Contents & Storage
2°C to 25°C