Ethanol Denatured Alcohol 70%, LabServ Pronalys
LabServ Pronalys™

Ethanol Denatured Alcohol 70%, LabServ Pronalys

Ethanol denatured alcohol 70% 6x 500mL sterile filtered through 0.22um trigger spray individually bagged MPI Approved C44
Have Questions?
Catalog number BSPSTE70-R
also known as BSPSTE70/R
Price (USD)
-
Ethanol denatured alcohol 70% 6x 500mL sterile filtered through 0.22um trigger spray individually bagged MPI Approved C44
  • Ethanol denatured alcohol 70% 6x 500mL sterile filtered through 0.22um trigger spray individually bagged MPI Approved C44 No. 64-17-5 Plastic Bottle
Specifications
Boiling Point78°C
CAS64-17-5
Chemical Name or MaterialEthyl alcohol
Melting Point-114°C
PackagingPlastic
GradeLaboratory
Percent Purity70%
Quantity500 mL
Unit SizeEach