Isopropyl Alcohol IPA 70%, LabServ Pronalys™
Isopropyl Alcohol IPA 70%, LabServ Pronalys™
LabServ Pronalys™

Isopropyl Alcohol IPA 70%, LabServ Pronalys™

Isopropyl alcohol IPA 70% 6x 500mL sterile filtered through 0.22um trigger spray individually bagged MPI Approved C44
Have Questions?
Catalog number BSPSTI70
Price (USD)
-
Isopropyl alcohol IPA 70% 6x 500mL sterile filtered through 0.22um trigger spray individually bagged MPI Approved C44
  • Isopropyl alcohol IPA 70% 6x 500mL sterile filtered through 0.22um trigger spray individually bagged MPI Approved C44 Plasti bottle CAS No. 67-63-0
Specifications
Boiling Point82°C
CAS67-63-0
Chemical Name or MaterialIsopropyl Alcohol [for Spectrophotometry]
IUPAC Namepropan-2-ol
InChI KeyKFZMGEQAYNKOFK-UHFFFAOYSA-N
Melting Point-89.5°C
Molecular FormulaC3H8O
PackagingPlastic
SMILESCC(C)O
GradeLaboratory
Molecular Weight (g/mol)60.10
Percent Purity70%
Quantity500 mL
Unit SizeEach