Rose-Bengal Chloramphenicol Agar Base (Dehydrated)
Rose-Bengal Chloramphenicol Agar Base (Dehydrated)
Thermo Scientific™
Rose-Bengal Chloramphenicol Agar Base (Dehydrated)
Isolate and enumerate yeasts and molds from a wide variety of foodstuffs with this selective medium.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
CM0549T5 kg
CM0549B500 g
CM0549R2.5 kg
Catalog number CM0549T
Price (USD)
-
Request A Quote
Quantity:
5 kg
Isolate and enumerate yeasts and molds from a wide variety of foodstuffs with Thermo Scientific™ Oxoid™ Rose Bengal Chloramphenicol Agar Base (Dehydrated). The neutral pH of the medium and the presence of selective agent chloramphenicol helps to suppress the bacterial growth, allowing molds and yeasts to grow. Rose Bengal in the medium is taken up by mold and yeast colonies assisting the enumeration of small colonies1. Rose Bengal also controls the size and height of mold colonies preventing the overgrowth, resulting in improved enumeration and detection.
Use Rose Bengal Chloramphenicol Agar for selective enumeration of molds and yeast from foods
  • Easy colony acounting: Rose-Bengal in the medium controls the size and height of mold colonies preventing overgrowth and making colony counting easier.
  • Selective: appropriate for isolation of yeasts and molds from foods whose associated flora consists mainly of Gram-negative rod-shaped bacteria
  • Suitable for higher and prolonged incubation temperatures.
The choice of a suitable medium for enumeration of yeasts and molds is greatly dependent on the nature of the foodstuffs under investigation and the organisms that occur on them2. Rose-Bengal Chloramphenicol Agar is recommended for fresh proteinaceous foods whose associated flora consists mainly of Gram-negative rod-shaped bacteria, although it should be noted that chloramphenicol alone may not be sufficient to inhibit the bacterial background. Because of the stability of chloramphenicol, Rose-Bengal Chloramphenicol Agar is also suitable when higher and prolonged incubation temperatures around 35°C are required.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Specifications
Quantity5 kg
FormPowder
Product TypeAgar
Unit SizeEach