You need to enable JavaScript to run this app.
Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Home
▸
Shop All Products
▸
Tubing
▸
Vacuum Tubing
▸
Masterflex™ Norprene Tubing (A60 G)
Actual product may vary
Cole-Parmer™
Masterflex™ Norprene Tubing (A60 G)
Nonconductive, heat-sealable, nonaging, nonoxidizing, with superior acid and alkali resistance. Opaque black.
Have Questions?
Change view
Catalog Number
Tubing Size
Flow Rate
CP0640417
also known as CP-CP0640417-EACH
17
1700mL/min.
CP0640418
also known as CP-CP0640418-EACH
18
2300mL/min.
2 Options
Catalog number
CP0640417
also known as CP-CP0640417-EACH
Price (USD)
-
Tubing Size:
17
Flow Rate:
1700mL/min.
Nonconductive, heat-sealable, nonaging, nonoxidizing, with superior acid and alkali resistance. Opaque black
Applications:
For applications requiring excellent chemical, heat, ozone, and ultraviolet (UV) light resistance.
Characteristics:
Nonconductive, heat-sealable, nonaging, nonoxidizing, with superior acid and alkali resistance. Opaque black.
Specifications
Flow Rate
1700mL/min.
Hose Barb
1/4
Tubing Size
17
Color
Opaque Black
Diameter (English) Outer
0.663 in.
Diameter (Metric) Outer
17 mm
Material
Norprene
Overall Length (English)
50 ft.
Overall Length (Metric)
15.24 m
Product Type
Tubing
Unit Size
Each