Masterflex™ L/S, Chem-Durance Bio Tubing
Actual product may vary
Cole-Parmer™
Masterflex™ L/S, Chem-Durance Bio Tubing
Excellent chemical resistance and long pumping life!
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTubing SizeFlow Rate
CP0644217
also known as CP-CP0644217-EACH
171700mL/min.
CP0644213
also known as CP-CP0644213-EACH
1336mL/min.
CP0644214
also known as CP-CP0644214-EACH
14130mL/min.
CP0644216
also known as CP-CP0644216-EACH
16480mL/min.
CP0644218
also known as CP-CP0644218-EACH
182300mL/min.
CP0644225
also known as CP-CP0644225-EACH
251000mL/min.
Catalog number CP0644217
also known as CP-CP0644217-EACH
Price (USD)
-
Tubing Size:
17
Flow Rate:
1700mL/min.
Excellent chemical resistance and long pumping life!
Excellent chemical resistance and long pumping life!
  • Now meets USP Class VI requirements!
  • Flexible outer jacket acts as an insulating layer to absorb and dissipate heat from the rollers — extending the life of the tubing
  • Plasticizer-free, smooth bore liner is hydrophobic and resists the absorption/adsorption of aqueous fluids
Specifications
FlexibilityFlexible
Flow Rate1700mL/min.
Hose Barb1/4
Tubing Size17
Diameter (English) Outer0.66 in.
Diameter (Metric) Outer17 mm
Product TypePump Tubing
Unit SizeEach