You need to enable JavaScript to run this app.
Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Masterflex™ PharMed BPT Tubing
Cole-Parmer™
Masterflex™ PharMed BPT Tubing
Cell and tissue culture work and pharmaceutical uses and good for light-sensitive samples.
Have Questions?
Change view
Catalog Number
Tubing Size
Flow Rate
CP0650836
also known as CP-CP0650836-EACH
36
3400mL/min.
CP0650813
also known as CP-CP0650813-EACH
13
36mL/min.
CP0650835
also known as CP-CP0650835-EACH
35
2600mL/min.
3 Options
Catalog number
CP0650836
also known as CP-CP0650836-EACH
Price (USD)
-
Tubing Size:
36
Flow Rate:
3400mL/min.
Cell and tissue culture work and pharmaceutical uses and good for light-sensitive samples.
Applications:
Cell and tissue culture work and pharmaceutical uses. Also good for light-sensitive samples.
Characteristics:
Nontoxic; nonhemolytic; extremely low permeability; resists acids, alkalies, oxidizing agents, and animal and vegetable oils; heat-sealable, bondable, and formable. Opaque beige.
Certifications:
FDA compliant and meets USP Class VI, EP, and NSF-51 requirements. Tubing is manufactured according to GMP.
Temperature range:
–60 to 270°F (–51 to 132°C)
Sterilize:
Autoclave, ethylene oxide, or gamma radiation.
Specifications
Flow Rate
3400mL/min.
Hose Barb
3/8
Tubing Size
36
Color
Opaque Beige
Diameter (English) Outer
1.41 in.
Diameter (Metric) Outer
36 mm
Product Type
Pump Tubing
Unit Size
Each