Masterflex™ Peroxide-cured Silicone Tubing, I/P 82, 25  ft.
Cole-Parmer™
Masterflex™ Peroxide-cured Silicone Tubing, I/P 82, 25 ft.
Excellent heat resistance, low compression set, odorless, nontoxic. Translucent.
Have Questions?
Catalog NumberTubing Size
CP9640082
also known as CP-CP9640082-EACH
82
Catalog number CP9640082
also known as CP-CP9640082-EACH
Price (USD)
-
Tubing Size:
82
Excellent heat resistance, low compression set, odorless, nontoxic. Translucent.
  • Applications: Pharmaceutical, biotechnology, peristaltic pumping, and food applications.
  • Characteristics: Excellent heat resistance, low compression set, odorless, nontoxic. Translucent.
Specifications
Flow Rate13000mL/min.
Hose Barb1/2
Tubing Size82
ColorTranslucent
Diameter (English) Outer3.22 in.
Diameter (Metric) Outer82 mm
MaterialSilicone
Product TypeTubing
Unit SizeEach