Disodium phosphate, Dominion Chemicals
Dominion Chemicals

Disodium phosphate, Dominion Chemicals

Disodium phosphate food grade
Have Questions?
Catalog number DSP001
Price (USD)
-
Disodium phosphate food grade
  • Disodium phosphate
  • Food grade
  • CAS No. 7558-79-4
Specifications
CAS7558-79-4
Chemical Name or MaterialDisodium phosphate
IUPAC Namedisodium hydrogen phosphate
InChI KeyBNIILDVGGAEEIG-UHFFFAOYSA-L
MDL NumberMFCD00003496
Melting Point243°C to 245°C
Molecular FormulaHNa2O4P
PackagingPlastic
SMILES[Na+].[Na+].OP([O-])([O-])=O
Molecular Weight (g/mol)141.96
Quantity25 kg
Unit SizeEach