AlbuMax II 20% Solution BSA, IRR, 1 L
Gibco™
AlbuMax II 20% Solution BSA, IRR, 1 L
Lipid-rich bovine serum albumin for cell culture. It is chromatographically purified and has an IgG content ≤1.0%. 20% solution with additional irradiation.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
E003001PR5 L
E003001PK1 L
Catalog number E003001PR
Price (USD)
-
Quantity:
5 L
Lipid-rich bovine serum albumin for cell culture. It is chromatographically purified and has an IgG content ≤1.0%. 20% solution with additional irradiation.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
Cell TypeMammalian Cells
Product TypeBSA
Quantity5 L
FormLiquid
Unit SizeEach