Modifying Enzymes
Thermo Scientific™

Modifying Enzymes

Thermo Scientific enzymes for in vitro transcription are designed for efficient and reliable RNA synthesis. These enzymes cover the entire synthesis workflow from template preparation to post-transcriptional modifications, ensuring optimal performance and high yields of synthesized RNA.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityConcentration
EL002AF1500 U30 U/μL
EM167AF20,000 U50 U/μL
EO166AF4000 U10 U/μL
4387661AF1000 U10 U/μL
EN230AF5000 U50 U/μL
Catalog number EL002AF
Price (USD)
-
Quantity:
1500 U
Concentration:
30 U/μL

Thermo Scientific enzymes for in vitro transcription are designed for efficient and reliable RNA synthesis. These enzymes cover the entire synthesis workflow from template preparation to post-transcriptional modifications, ensuring optimal performance and high yields of synthesized RNA.

High quality RNA synthesized by in vitro transcription is essential for RNA research and this versatile technique can be used in a variety of applications including the introduction of modified nucleotides into RNA transcripts for enhanced translational capacity and reduced immune activity of the cells.

Thermo Scientific enzymes for in vitro transcription are designed for efficient and reliable RNA synthesis. These enzymes cover the entire synthesis workflow from template preparation to post-transcriptional modifications, ensuring optimal performance and high yields of synthesized RNA.

High quality RNA synthesized by in vitro transcription is essential for RNA research and this versatile technique can be used in a variety of applications including the introduction of modified nucleotides into RNA transcripts for enhanced translational capacity and reduced immune activity of the cells.

Features

  • Convenient stand-alone enzymes and buffers for workflow optimization
  • No animal-origin components in final product composition
  • Reliable and reproducible results with rigorous quality testing
  • Scalable and available as OEM products

Applications

  • CRISPR/Cas-based genome editing
  • Gene expression
  • Generation of pluripotent cells
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Compatible BufferT4 RNA Ligase Reaction Buffer
For Use With (Application)RNA Ligation
Inactivation Temperature70°C
Product TypeT4 RNA Ligase
Quantity1500 U
Concentration30 U/μL
FormLiquid
Research CategoryRNA Research
Sensitive to Heat InactivationYes
Unit SizeEach
Contents & Storage
30 U/μL T4 RNA Ligase, 50 μL

Store at -20°C.