Hexanes, Certified AR for Analysis, 95% N-Hexane Approx, Fisher Chemical™
Hexanes, Certified AR for Analysis, 95% N-Hexane Approx, Fisher Chemical™
Hexanes, Certified AR for Analysis, 95% N-Hexane Approx, Fisher Chemical™
Fisher Chemical™

Hexanes, Certified AR for Analysis, 95% N-Hexane Approx, Fisher Chemical™

C6H14, CAS Number-92112-69-1, hexanes, dipropyl, gettysolve-b, skellysolve b, esani, n-hexane, heksan, hexyl hydride, hexanen, hexan, 5L, 69 deg.C, CHEBI:29021, Colorless, 86.18g/mol, VLKZOEOYAKHREP-UHFFFAOYSA-N, -95 deg.C, 9520, 86.178, Metal can, Liquid, 8058, CCCCCC, 2.97, 160mbar at 20 deg.C
Have Questions?
Catalog NumberQuantityPackaging
H-0355-21
also known as H/0355/21
5 LMetal Can
Catalog number H-0355-21
also known as H/0355/21
Price (USD)
-
Quantity:
5 L
Packaging:
Metal Can
Request bulk or custom format
Specifications
Boiling Point69°C
CAS92112-69-1
ChEBICHEBI:29021
ColorColorless
Formula Weight86.18g/mol
View more
C6H14, CAS Number-92112-69-1, hexanes, dipropyl, gettysolve-b, skellysolve b, esani, n-hexane, heksan, hexyl hydride, hexanen, hexan, 5L, 69 deg.C, CHEBI:29021, Colorless, 86.18g/mol, VLKZOEOYAKHREP-UHFFFAOYSA-N, -95 deg.C, 9520, 86.178, Metal can, Liquid, 8058, CCCCCC, 2.97, 160mbar at 20 deg.C