Thermo Scientific Chemicals

2-Fluoroisobutyric acid, 95%, Thermo Scientific Chemicals

Catalog number: H25837.MD
250 mg, Each
Thermo Scientific Chemicals

2-Fluoroisobutyric acid, 95%, Thermo Scientific Chemicals

Catalog number: H25837.MD
250 mg, Each
Quantity
Catalog number: H25837.MD
also known as H25837-MD
Price (USD)

Chemical Identifiers

CAS
63812-15-7
IUPAC Name
2-fluoro-2-methylpropanoic acid
Molecular Formula
C4H7FO2
InChI Key
NZDOWZQRNZLBOY-UHFFFAOYSA-N
SMILES
CC(C)(F)C(O)=O
Appearance (Color)
Clear colorless to pale yellow to pale brown
Assay (Aqueous acid-base Titration)
≥94.0 to ≤106.0%
Refractive Index
1.3820-1.3880 @ 20?C
Form
Liquid

Description

2-Fluoroisobutyric acid chloride was synthesized from 2-fluoroisobutyric acid and oxalyl chloride. Reactant in the synthesis of 2-Fluoro-Acids, Esters, and amides From a-Dicyanoepoxides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Fluoroisobutyric acid chloride was synthesized from 2-fluoroisobutyric acid and oxalyl chloride. Reactant in the synthesis of 2-Fluoro-Acids, Esters, and amides From a-Dicyanoepoxides.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text