Thermo Scientific Chemicals

Triethylsulfonium bis(trifluoromethylsulfonyl)imide, 98%, Thermo Scientific Chemicals

Catalog number: H27116.06
5 g, Each
Thermo Scientific Chemicals

Triethylsulfonium bis(trifluoromethylsulfonyl)imide, 98%, Thermo Scientific Chemicals

Catalog number: H27116.06
5 g, Each
Quantity
Catalog number: H27116.06
also known as H27116-06
Price (USD)

Chemical Identifiers

CAS
321746-49-0
IUPAC Name
triethylsulfanium; trifluoro[(trifluoromethanesulfonylazanidyl)sulfonyl]methane
Molecular Formula
C8H15F6NO4S3
InChI Key
BLODSRKENWXTLO-UHFFFAOYSA-N
SMILES
CC[S+](CC)CC.FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F
Appearance (Color)
Clear colorless to yellow
Assay by NMR
≥97.5%
Assay from Supplier's CofA
≥97.5%
Water Content (Karl Fischer Titration)
≤0.5%
Proton NMR
Conforms to structure

Description

Proton-Conducting Properties of a Bronsted Acid-Base Ionic Liquid and Ionic Melts Consisting of Bis(trifluoromethanesulfonyl)imide and Benzimidazole for Fuel Cell Electrolytes. Symmetric and aliphatic trialkylsulfonium cation form room temperature molten salts (RTMS) with bis(trifluoromethylsulfonyl)imide (TFSI).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Proton-Conducting Properties of a Bronsted Acid-Base Ionic Liquid and Ionic Melts Consisting of Bis(trifluoromethanesulfonyl)imide and Benzimidazole for Fuel Cell Electrolytes. Symmetric and aliphatic trialkylsulfonium cation form room temperature molten salts (RTMS) with bis(trifluoromethylsulfonyl)imide (TFSI).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text