Thermo Scientific Chemicals

1-Amino-1-cyclopropanecarboxylic acid hydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: H27264.03
1 g, Each
Thermo Scientific Chemicals

1-Amino-1-cyclopropanecarboxylic acid hydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: H27264.03
1 g, Each
Quantity
Catalog number: H27264.03
also known as H27264-03
Price (USD)

Chemical Identifiers

CAS
68781-13-5
IUPAC Name
1-aminocyclopropane-1-carboxylic acid hydrochloride
Molecular Formula
C4H8ClNO2
InChI Key
WTZRHZRMLYNBRM-UHFFFAOYSA-N
SMILES
Cl.NC1(CC1)C(O)=O
Appearance (Color)
White to pale cream
Assay (Titration ex Chloride)
≥96.0 to ≤104.0%
Form
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text