4-Amino-2-methoxybenzoic acid, 97%
4-Amino-2-methoxybenzoic acid, 97%
4-Amino-2-methoxybenzoic acid, 97%
Thermo Scientific Chemicals

4-Amino-2-methoxybenzoic acid, 97%

CAS: 2486-80-8 | C8H9NO3 | 167.164 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H27419.06
also known as H27419-06
Price (USD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS2486-80-8
IUPAC Name4-amino-2-methoxybenzoic acid
Molecular FormulaC8H9NO3
InChI KeyOLJXRTRRJSMURJ-UHFFFAOYSA-N
SMILESCOC1=CC(N)=CC=C1C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (Silylated GC)≥96.0%
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
Appearance (Color)White to cream
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only