Thermo Scientific Chemicals

3-Iodobenzeneboronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Catalog number: H28115.03
1 g, Each
Thermo Scientific Chemicals

3-Iodobenzeneboronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Catalog number: H28115.03
1 g, Each
Quantity
Catalog number: H28115.03
also known as H28115-03
Price (USD)

Chemical Identifiers

CAS
408492-28-4
IUPAC Name
2-(3-iodophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Formula
C12H16BIO2
InChI Key
HSHFNMSHDHAHDN-UHFFFAOYSA-N
SMILES
CC1(C)OB(OC1(C)C)C1=CC(I)=CC=C1
Form
Powder
Appearance (Color)
Cream
Assay (GC)
≥96.0%
Melting Point (clear melt)
67.0-76.0?C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text