3-Methylbenzoic anhydride, 97%
3-Methylbenzoic anhydride, 97%
3-Methylbenzoic anhydride, 97%
Thermo Scientific Chemicals

3-Methylbenzoic anhydride, 97%

CAS: 21436-44-2 | C16H14O3 | 254.285 g/mol
Have Questions?
Catalog NumberQuantity
H32739.06
also known as H32739-06
5 g
Catalog number H32739.06
also known as H32739-06
Price (USD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS21436-44-2
IUPAC Name3-methylbenzoyl 3-methylbenzoate
Molecular FormulaC16H14O3
InChI KeyDGZSQLHPEZFGSI-UHFFFAOYSA-N
SMILESCC1=CC(=CC=C1)C(=O)OC(=O)C1=CC(C)=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (GC)≥96.0%
FormCrystals or powder or crystalline powder or flakes
Melting Point (clear melt)65.5-74.5°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only