3-Amino-5-bromo-1-methyl-1H-indazole, 97%
3-Amino-5-bromo-1-methyl-1H-indazole, 97%
3-Amino-5-bromo-1-methyl-1H-indazole, 97%
Thermo Scientific Chemicals

3-Amino-5-bromo-1-methyl-1H-indazole, 97%

CAS: 1000018-06-3 | C8H8BrN3 | 226.077 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Catalog number H33202.03
also known as H33202-03
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS1000018-06-3
IUPAC Name5-bromo-1-methyl-1H-indazol-3-amine
Molecular FormulaC8H8BrN3
InChI KeyBKBSBRJIGMVBFM-UHFFFAOYSA-N
SMILESCN1N=C(N)C2=CC(Br)=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale cream
Assay (HPLC)≥96.0%
Melting Point (clear melt)130.0-140.0?C
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only