Thermo Scientific Chemicals

Methyl 2-hydroxynicotinate, 97%, Thermo Scientific Chemicals

Catalog number: H33334.03
1 g, Each
Thermo Scientific Chemicals

Methyl 2-hydroxynicotinate, 97%, Thermo Scientific Chemicals

Catalog number: H33334.03
1 g, Each
Quantity
Catalog number: H33334.03
also known as H33334-03
Price (USD)

Chemical Identifiers

CAS
10128-91-3
IUPAC Name
methyl 2-oxo-1,2-dihydropyridine-3-carboxylate
Molecular Formula
C7H7NO3
InChI Key
SILBTMNCGYLTOK-UHFFFAOYSA-N
SMILES
COC(=O)C1=CC=CNC1=O
Appearance (Color)
White to cream
Assay (HPLC)
≥96.0%
Loss on Drying
≤1.5% (105?C/constant weight)
Form
Powder
Identification (FTIR)
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text