Ethyl 2-amino-5,6,7,8-tetrahydro-4H-cyclohepta[b]thiophene-3-carboxylate, 96%
Ethyl 2-amino-5,6,7,8-tetrahydro-4H-cyclohepta[b]thiophene-3-carboxylate, 96%
Ethyl 2-amino-5,6,7,8-tetrahydro-4H-cyclohepta[b]thiophene-3-carboxylate, 96%
Thermo Scientific Chemicals

Ethyl 2-amino-5,6,7,8-tetrahydro-4H-cyclohepta[b]thiophene-3-carboxylate, 96%

CAS: 40106-13-6 | C12H17NO2S | 239.333 g/mol
Have Questions?
Catalog NumberQuantity
H34116.06
also known as H34116-06
5 g
Catalog number H34116.06
also known as H34116-06
Price (USD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS40106-13-6
IUPAC Nameethyl 2-amino-4H,5H,6H,7H,8H-cyclohepta[b]thiophene-3-carboxylate
Molecular FormulaC12H17NO2S
InChI KeyXTUHIGALMIGZST-UHFFFAOYSA-N
SMILESCCOC(=O)C1=C(N)SC2=C1CCCCC2
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Melting Point (clear melt)85-91?C
Proton NMRConforms to structure
Assay (GC)≥95.0%
Appearance (Color)White to pale cream or pale yellow to pale orange
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Soluble in methanol and ethanol.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only
K. A. Nirmala; Vasu; D. Chopra; S. Mohan and M. R. Prasad. Ethyl 2-amino-5,6,7,8-tetra­hydro-4H-cyclo­hepta­[b]­thio­phene-3-carboxyl­ate. Acta Cryst. 1999, E61 o1541-o1543.