1-Methyl-3-n-octylimidazolium bis(trifluoromethylsulfonyl)imide, 99%
1-Methyl-3-n-octylimidazolium bis(trifluoromethylsulfonyl)imide, 99%
1-Methyl-3-n-octylimidazolium bis(trifluoromethylsulfonyl)imide, 99%
Thermo Scientific Chemicals

1-Methyl-3-n-octylimidazolium bis(trifluoromethylsulfonyl)imide, 99%

CAS: 178631-04-4 | C14H23F6N3O4S2 | 475.465 g/mol
Have Questions?
Catalog NumberQuantity
H35352.14
also known as H35352-14
25 g
Catalog number H35352.14
also known as H35352-14
Price (USD)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS178631-04-4
IUPAC Name3-methyl-1-octyl-1H-imidazol-3-ium; trifluoro[(trifluoromethanesulfonylazanidyl)sulfonyl]methane
Molecular FormulaC14H23F6N3O4S2
InChI KeyLECQXINNQGHJBM-UHFFFAOYSA-N
SMILESCCCCCCCCN1C=C[N+](C)=C1.FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
FormViscous liquid
Identification (FTIR)Conforms
Residual halide<100ppm
Ion Chromatography≥98.5%
Water Content (Karl Fischer Titration)<100ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only