Benzo[b]thiophene-3-carboxylic acid, 96%
Benzo[b]thiophene-3-carboxylic acid, 96%
Benzo[b]thiophene-3-carboxylic acid, 96%
Thermo Scientific Chemicals

Benzo[b]thiophene-3-carboxylic acid, 96%

CAS: 5381-25-9 | C9H6O2S | 178.205 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number H50105.14
also known as H50105-14
Price (USD)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS5381-25-9
IUPAC Name1-benzothiophene-3-carboxylic acid
Molecular FormulaC9H6O2S
InChI KeyDRBLTQNCQJXSNU-UHFFFAOYSA-N
SMILESOC(=O)C1=CSC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Light yellow
FormPowder
Assay (HPLC)>95.0%
Benzo[b]thiophene-3-carboxylic acid is used as organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzo[b]thiophene-3-carboxylic acid is used as organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only
Peter E.Cross; Roger P.Dickinson; M.John Parry; Michael J.Randall. Selective thromboxane synthetase inhibitors. 3. 1H-Imidazol-1-yl-substituted benzo[b]furan-, benzo[b]thiophene- and indole-2- and 3-carboxylic acids. J. Med. Chem. 1986, 29, (9),1637-1643