2-(Trifluoromethyl)pyrimidine-4-carboxamide, 97%, Thermo Scientific Chemicals
2-(Trifluoromethyl)pyrimidine-4-carboxamide, 97%, Thermo Scientific Chemicals
2-(Trifluoromethyl)pyrimidine-4-carboxamide, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-(Trifluoromethyl)pyrimidine-4-carboxamide, 97%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
H52803.03
also known as H52803-03
1 g
Catalog number H52803.03
also known as H52803-03
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Specifications
CAS914348-10-0
Chemical Name or Material2-(Trifluoromethyl)pyrimidine-4-carboxamide
Formula Weight191.11
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
2-(Trifluoromethyl)pyrimidine-4-carboxamide is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Trifluoromethyl)pyrimidine-4-carboxamide is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only