Thermo Scientific Chemicals

Pyrimidine-5-carboxaldehyde, 97%, Thermo Scientific Chemicals

Catalog number: H54448.06
5 g, Each
Thermo Scientific Chemicals

Pyrimidine-5-carboxaldehyde, 97%, Thermo Scientific Chemicals

Catalog number: H54448.06
5 g, Each
Quantity
Catalog number: H54448.06
also known as H54448-06
Price (USD)

Chemical Identifiers

CAS
10070-92-5
IUPAC Name
pyrimidine-5-carbaldehyde
Molecular Formula
C5H4N2O
InChI Key
FREJAOSUHFGDBW-UHFFFAOYSA-N
SMILES
O=CC1=CN=CN=C1
Form
Powder
Assay (unspecified)
>96.0%
Appearance (Color)
Yellow

Description

Pyrimidine-5-carboxaldehyde is used in the preparation of (S)- and (R)-pyrimidyl alkanol by reacting with diisopropylzinc and (n-butyl)ethyl(n-hexyl)(n-propyl)methane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pyrimidine-5-carboxaldehyde is used in the preparation of (S)- and (R)-pyrimidyl alkanol by reacting with diisopropylzinc and (n-butyl)ethyl(n-hexyl)(n-propyl)methane.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text