Thermo Scientific Chemicals

3-Methoxyphenylacetylene, 96%, Thermo Scientific Chemicals

Catalog number: H55254.MD
250 mg, Each
Thermo Scientific Chemicals

3-Methoxyphenylacetylene, 96%, Thermo Scientific Chemicals

Catalog number: H55254.MD
250 mg, Each
Quantity
Catalog number: H55254.MD
also known as H55254-MD
Price (USD)

Chemical Identifiers

CAS
768-70-7
IUPAC Name
1-ethynyl-3-methoxybenzene
Molecular Formula
C9H8O
InChI Key
ZASXCTCNZKFDTP-UHFFFAOYSA-N
SMILES
COC1=CC=CC(=C1)C#C
Assay (HPLC)
>95.0%

Description

It is a useful aromatic compound.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a useful aromatic compound.

Solubility
Slightly soluble in water.

Notes
Storage: Keep container tightly closed. Store in a cool, dark and well-ventilated place and away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text