Thermo Scientific Chemicals

3-n-Hexylthiophene, 98%, Thermo Scientific Chemicals

Catalog number: H55891.06
5 g, Each
Thermo Scientific Chemicals

3-n-Hexylthiophene, 98%, Thermo Scientific Chemicals

Catalog number: H55891.06
5 g, Each
Quantity
Catalog number: H55891.06
also known as H55891-06
Price (USD)

Chemical Identifiers

CAS
1693-86-3
IUPAC Name
3-hexylthiophene
Molecular Formula
C10H16S
InChI Key
JEDHEMYZURJGRQ-UHFFFAOYSA-N
SMILES
CCCCCCC1=CSC=C1
Appearance (Color)
Colorless to pale yellow
Assay (GC)
≥97.5%
Infrared spectrum
Conforms to structure
Form
Liquid

Description

3-n-Hexylthiophene is a conducting polymer precursor. may be used in the synthesis of poly(3-hexylthiophene). MgKR X-ray and low energy electron induced oligomerization of physisorbed layers of 3-hexylthiophene condensed on clean gold results in the formation of an organic film.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-n-Hexylthiophene is a conducting polymer precursor. may be used in the synthesis of poly(3-hexylthiophene). MgKR X-ray and low energy electron induced oligomerization of physisorbed layers of 3-hexylthiophene condensed on clean gold results in the formation of an organic film.

Solubility
Partly soluble in water.

Notes
Store at room temperature. Avoid strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text