Thermo Scientific Chemicals

Methyl thiophene-3-carboxylate, 97%, Thermo Scientific Chemicals

Catalog number: H61488.06
5 g, Each
Thermo Scientific Chemicals

Methyl thiophene-3-carboxylate, 97%, Thermo Scientific Chemicals

Catalog number: H61488.06
5 g, Each
Quantity
Catalog number: H61488.06
also known as H61488-06
Price (USD)

Chemical Identifiers

CAS
22913-26-4
IUPAC Name
methyl thiophene-3-carboxylate
Molecular Formula
C6H6O2S
InChI Key
ZTRAEMILTFNZSM-UHFFFAOYSA-N
SMILES
COC(=O)C1=CSC=C1
Assay (GC)
>96.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text