3-Chloro-4-cyanobenzeneboronic acid, 96%
3-Chloro-4-cyanobenzeneboronic acid, 96%
3-Chloro-4-cyanobenzeneboronic acid, 96%
Thermo Scientific Chemicals

3-Chloro-4-cyanobenzeneboronic acid, 96%

CAS: 1008415-02-8 | C7H5BClNO2 | 181.382 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H61543.03
also known as H61543-03
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Specifications
CAS1008415-02-8
Chemical Name or Material3-Chloro-4-cyanobenzeneboronic acid
Formula Weight164.93
Health Hazard 1H302-H315-H319-H335
Health Hazard 3P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only