EH-IDTBr, Thermo Scientific Chemicals
EH-IDTBr, Thermo Scientific Chemicals
EH-IDTBr, Thermo Scientific Chemicals
Thermo Scientific Chemicals

EH-IDTBr, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
250 mg
500 mg
Catalog number H66142.ME
also known as H66142-ME
Price (USD)
-
Quantity:
500 mg
Request bulk or custom format
Chemical Identifiers
CAS2102510-60-9
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Black
Bandgap energy1.64 ? 0.02 eV
FormFlake or threadlike
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only